Merck
  • Timothy syndrome is associated with activity-dependent dendritic retraction in rodent and human neurons.
Timothy syndrome is associated with activity-dependent dendritic retraction in rodent and human neurons.
Nature neuroscience (2013-01-15)
Jocelyn F Krey, Sergiu P Paşca, Aleksandr Shcheglovitov, Masayuki Yazawa, Rachel Schwemberger, Randall Rasmusson, Ricardo E Dolmetsch
ABSTRACT
L-type voltage gated calcium channels have an important role in neuronal development by promoting dendritic growth and arborization. A point mutation in the gene encoding Ca(V)1.2 causes Timothy syndrome, a neurodevelopmental disorder associated with autism spectrum disorders (ASDs). We report that channels with the Timothy syndrome alteration cause activity-dependent dendrite retraction in rat and mouse neurons and in induced pluripotent stem cell (iPSC)-derived neurons from individuals with Timothy syndrome. Dendrite retraction was independent of calcium permeation through the mutant channel, was associated with ectopic activation of RhoA and was inhibited by overexpression of the channel-associated GTPase Gem. These results suggest that Ca(V)1.2 can activate RhoA signaling independently of Ca(2+) and provide insights into the cellular basis of Timothy syndrome and other ASDs.
Sigma-Aldrich
Anti-Calcium Channel Antibody, Voltage Gated α 1C, Chemicon®, from rabbit
Pricing and availability is not currently available.
Roche
Anti-HA-Biotin, High Affinity (3F10), from rat IgG1
Pricing and availability is not currently available.