Merck
  • OmpA-mediated rickettsial adherence to and invasion of human endothelial cells is dependent upon interaction with α2β1 integrin.
OmpA-mediated rickettsial adherence to and invasion of human endothelial cells is dependent upon interaction with α2β1 integrin.
Cellular microbiology (2012-11-14)
Robert D Hillman, Yasmine M Baktash, Juan J Martinez
ABSTRACT
Rickettsia conorii, a member of the spotted fever group (SFG) of the genus Rickettsia and causative agent of Mediterranean spotted fever, is an obligate intracellular pathogen capable of infecting various mammalian cell types. SFG rickettsiae express two major immunodominant surface cell antigen (Sca) proteins, OmpB (Sca5) and OmpA (Sca0). While OmpB-mediated entry has been characterized, the contribution of OmpA has not been well defined. Here we show OmpA expression in Escherichia coli is sufficient to mediate adherence to and invasion of non-phagocytic human endothelial cells. A recombinant soluble C-terminal OmpA protein domain (954-1735) with predicted structural homology to the Bordetella pertussis pertactin protein binds mammalian cells and perturbs R. conorii invasion by interacting with several mammalian proteins including β1 integrin. Using functional blocking antibodies, small interfering RNA transfection, and mouse embryonic fibroblast cell lines, we illustrate the contribution of α2β1 integrin as a mammalian ligand involved in R. conorii invasion of primary endothelial cells. We further demonstrate that OmpA-mediated attachment to mammalian cells is in part dependent on a conserved non-continuous RGD motif present in a predicted C-terminal 'pertactin' domain in OmpA.Our results demonstrate that multiple adhesin-receptor pairs are sufficient in mediating efficient bacterial invasion of R. conorii.
Sigma-Aldrich
β Integrin Screening Kit
Sigma-Aldrich
α Integrin Blocking and IHC Kit
Sigma-Aldrich
Anti-Integrin α2 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4G11, FITC conjugated, clone P4G11, Chemicon®, from mouse