Merck
  • A highly sensitive water-soluble system to sense glucose in aqueous solution.
A highly sensitive water-soluble system to sense glucose in aqueous solution.
Organic & biomolecular chemistry (2011-03-08)
Liheng Feng, Fei Liang, Yue Wang, Ming Xu, Xiaoju Wang
ABSTRACT
A glucose sensing switch is formed by water soluble conjugated polymer (PP-S-BINOL) and boronic acid-functionalized benzyl viologen (o-BBV). The two-component system shows a high sensitivity for glucose sensing with a 17-fold increase in the fluorescence intensity in the presence of 100 mM glucose.
Sigma-Aldrich
1,1′-Bi-2-naphthol, 99%