Merck
Sphingosine modulates interleukin-6 synthesis in osteoblasts.
Journal of cellular biochemistry (1998-08-26)
O Kozawa, H Tokuda, H Matsuno, T Uematsu
ABSTRACT
We previously reported that prostaglandin (PG)E1 and PGF2alpha induce the synthesis of interleukin-6 (IL-6) via activation of protein kinase (PK)A and PKC, respectively, in osteoblast-like MC3T3-E1 cells. In addition, we have shown that basic fibroblast growth factor (bFGF) elicits IL-6 synthesis through intracellular Ca2+ mobilization in these cells and that tumor necrosis factor-alpha (TNF) induces IL-6 synthesis through sphingosine 1-phosphate produced by sphingomyelin hydrolysis. In the present study, among sphingomyelin metabolites, we examined the effect of sphingosine on IL-6 synthesis induced by various agonists in MC3T3-E1 cells. Sphingosine inhibited the IL-6 synthesis induced by PGF2alpha or 12-O-tetradecanoylphorbol-13-acetate, an activator of PKC. Sphingosine suppressed the PGE1-induced IL-6 synthesis. The IL-6 synthesis induced by cholera toxin, forskolin, or dibutyryl cAMP was inhibited by sphingosine. Sphingosine inhibited the IL-6 synthesis induced by bFGF or A23187. However, sphingosine did not affect the IL-6 synthesis induced by interleukin-1. On the contrary, sphingosine enhanced the TNF-induced IL-6 synthesis. DL-threo-Dihydrosphingosine, an inhibitor of sphingosine kinase, reduced the enhancement by sphingosine as well as the TNF-effect. These results indicate that sphingosine modulates the IL-6 synthesis stimulated by various agonists in osteoblasts.