Merck
Substance P modulates colitis-associated fibrosis.
The American journal of pathology (2010-10-05)
Hon Wai Koon, David Shih, Iordanes Karagiannides, Dezheng Zhao, Zafeer Fazelbhoy, Tressia Hing, Hua Xu, Bao Lu, Norma Gerard, Charalabos Pothoulakis
ABSTRACT
Substance P (SP) and the neurokinin-1 receptor (NK-1R) are involved in the development of colitis and mucosal healing after colonic inflammation. We studied whether SP modulates colonic fibrosis by using a chronic model of trinitrobenzenesulfonic acid (TNBS)-induced colitis in wild-type (WT) and NK-1R-deficient (NK-1R KD) mice. We found increased mRNA expression levels of collagen, vimentin, and the fibrogenic factors transforming growth factor β1 and insulin-like growth factor 1 in the chronically inflamed colons of WT mice treated with repeated intracolonic TNBS administrations. Fibrosis in TNBS-treated mice was also evident immunohistochemically by collagen deposition in the colon. Treatment of TNBS-exposed WT mice with the NK-1R antagonist CJ-12255 reduced colonic inflammation, colonic fibrosis, fibroblast accumulation, and expression levels of the fibrogenic factors. NK-1R knockout mice chronically exposed to TNBS had similar colonic inflammation compared with WT, but reduced colonic fibrosis, fibroblast accumulation, and expression levels of fibrogenic factors. Immunohistochemical staining also showed co-localization of NK-1R with fibroblasts in inflamed colons of mice and in colonic mucosa of patients with Crohn's disease. Exposure of human colonic CCD-18Co fibroblasts to SP (10 nmol/L) increased cell migration. SP stimulated collagen synthesis in CCD-18Co fibroblasts in the presence of transforming growth factor β1 and insulin-like growth factor 1, and this effect was reduced by Akt inhibition. Thus, SP, via NK-1R, promotes intestinal fibrogenesis after chronic colitis by stimulating fibrotic responses in fibroblasts.
Sigma-Aldrich
Substance P, Neuropeptide found in high concentrations in the gut.
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), colorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with colorimetric detection.