Merck
  • Facile synthesis of nitrogen doped reduced graphene oxide as a superior metal-free catalyst for oxidation.
Facile synthesis of nitrogen doped reduced graphene oxide as a superior metal-free catalyst for oxidation.
Chemical communications (Cambridge, England) (2013-08-16)
Hongqi Sun, Yuxian Wang, Shizhen Liu, Lei Ge, Li Wang, Zhonghua Zhu, Shaobin Wang
ABSTRACT
Nitrogen (5.61 at%) doped reduced graphene oxide synthesized via a facile method was demonstrated as a superior metal-free catalyst for activation of peroxymonosulfate. Codoping with boron would further enhance the catalytic activity and the stability, providing a promising green material for environmental remediation.
Sigma-Aldrich
Ammonium pentaborate octahydrate
Pricing and availability is not currently available.