Merck
  • A Short Tandem Repeat-Enriched RNA Assembles a Nuclear Compartment to Control Alternative Splicing and Promote Cell Survival.
A Short Tandem Repeat-Enriched RNA Assembles a Nuclear Compartment to Control Alternative Splicing and Promote Cell Survival.
Molecular cell (2018-10-16)
Karen Yap, Svetlana Mukhina, Gen Zhang, Jason S C Tan, Hong Sheng Ong, Eugene V Makeyev
ABSTRACT
Functions of many long noncoding RNAs (lncRNAs) depend on their ability to interact with multiple copies of specific RNA-binding proteins (RBPs). Here, we devised a workflow combining bioinformatics and experimental validation steps to systematically identify RNAs capable of multivalent RBP recruitment. This uncovered a number of previously unknown transcripts encoding high-density RBP recognition arrays within genetically normal short tandem repeats. We show that a top-scoring hit in this screen, lncRNA PNCTR, contains hundreds of pyrimidine tract-binding protein (PTBP1)-specific motifs allowing it to sequester a substantial fraction of PTBP1 in a nuclear body called perinucleolar compartment. Importantly, PNCTR is markedly overexpressed in a variety of cancer cells and its downregulation is sufficient to induce programmed cell death at least in part by stimulating PTBP1 splicing regulation activity. This work expands our understanding of the repeat-containing fraction of the human genome and illuminates a novel mechanism driving malignant transformation of cancer cells.
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 18-39 human, ≥97% (HPLC)
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Anti-CUGBP1 Antibody, clone 3B1, clone 3B1, Upstate®, from mouse
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade