Merck
  • The LPA-LPA4 axis is required for establishment of bipolar morphology and radial migration of newborn cortical neurons.
The LPA-LPA4 axis is required for establishment of bipolar morphology and radial migration of newborn cortical neurons.
Development (Cambridge, England) (2018-09-16)
Nobuhiro Kurabayashi, Aiki Tanaka, Minh Dang Nguyen, Kamon Sanada
ABSTRACT
Newborn neurons in the developing neocortex undergo radial migration, a process that is coupled with their precise passage from multipolar to bipolar shape. The cell-extrinsic signals that govern this transition are, however, poorly understood. Here, we find that lysophosphatidic acid (LPA) signaling contributes to the establishment of a bipolar shape in mouse migratory neurons through LPA receptor 4 (LPA4). LPA4 is robustly expressed in migratory neurons. LPA4-depleted neurons show impaired multipolar-to-bipolar transition and become arrested in their migration. Further, LPA4-mediated LPA signaling promotes formation of the pia-directed process in primary neurons overlaid on neocortical slices. In addition, LPA4 depletion is coupled with altered actin organization as well as with destabilization of the F-actin-binding protein filamin A (FlnA). Finally, overexpression of FlnA rescues the morphology and migration defects of LPA4-depleted neurons. Thus, the LPA-LPA4 axis regulates bipolar morphogenesis and radial migration of newborn cortical neurons via remodeling of the actin cytoskeleton.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human LPAR4
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)