Merck
  • Selective laser melted titanium alloys for hip implant applications: Surface modification with new method of polymer grafting.
Selective laser melted titanium alloys for hip implant applications: Surface modification with new method of polymer grafting.
Journal of the mechanical behavior of biomedical materials (2018-08-14)
Subir Ghosh, Sylvester Abanteriba, Sherman Wong, Shadi Houshyar
ABSTRACT
A significant number of hip replacements (HR) fail permanently despite the success of the medical procedure, due to wear and progressive loss of osseointegration of implants. An ideal model should consist of materials with a high resistance to wear and with good biocompatibility. This study aims to develop a new method of grafting the surface of selective laser melted (SLM) titanium alloy (Ti-6Al-4V) with poly (2-methacryloyloxyethyl phosphorylcholine) (PMPC), to improve the surface properties and biocompatibility of the implant. PMPC was grafted onto the SLM fabricated Ti-6Al-4V, applying the following three techniques; ultraviolet (UV) irradiation, thermal heating both under normal atmosphere and UV irradiation under N2 gas atmosphere. Scanning electron microscopy (SEM), 3D optical profiler, energy-dispersive X-ray spectroscopy (EDS), X-ray photoelectron spectroscopy (XPS), and Fourier transform infrared spectroscopy (FTIR) were used to characterise the grafted surface. Results demonstrated that a continuous PMPC layer on the Ti-6Al-4V surface was achieved using the UV irradiation under N2 gas atmosphere technique, due to the elimination of oxygen from the system. As indicated in the results, one of the advantages of this technique is the presence of phosphorylcholine, mostly on the surface, which reveals the existence of a strong chemical bond between the grafted layer (PMPC) and substrate (Ti-6Al-4V). The nano-scratch test revealed that the PMPC grafted surface improves the mechanical strength of the surface and thus, protects the underlying implant substrate from scratching under high loads.
Sigma-Aldrich
Inhibitor removers, Prepacked column for removing hydroquinone and monomethyl ether hydroquinone
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methacryloyloxyethyl phosphorylcholine, contains ≤100 ppm MEHQ as inhibitor, 97%
Pricing and availability is not currently available.