Merck
  • Bioactive nanoengineered hydrogels for bone tissue engineering: a growth-factor-free approach.
Bioactive nanoengineered hydrogels for bone tissue engineering: a growth-factor-free approach.
ACS nano (2015-02-13)
Janet R Xavier, Teena Thakur, Prachi Desai, Manish K Jaiswal, Nick Sears, Elizabeth Cosgriff-Hernandez, Roland Kaunas, Akhilesh K Gaharwar
ABSTRACT
Despite bone's impressive ability to heal after traumatic injuries and fractures, a significant need still exists for developing strategies to promote healing of nonunion defects. To address this issue, we developed collagen-based hydrogels containing two-dimensional nanosilicates. Nanosilicates are ultrathin nanomaterials with a high degree of anisotropy and functionality that results in enhanced surface interactions with biological entities compared to their respective three-dimensional counterparts. The addition of nanosilicates resulted in a 4-fold increase in compressive modulus along with an increase in pore size compared to collagen-based hydrogels. In vitro evaluation indicated that the nanocomposite hydrogels are capable of promoting osteogenesis in the absence of any osteoinductive factors. A 3-fold increase in alkaline phosphatase activity and a 4-fold increase in the formation of a mineralized matrix were observed with the addition of the nanosilicates to the collagen-based hydrogels. Overall, these results demonstrate the multiple functions of nanosilicates conducive to the regeneration of bone in nonunion defects, including increased network stiffness and porosity, injectability, and enhanced mineralized matrix formation in a growth-factor-free microenvironment.
Sigma-Aldrich
TissueFab® - low endotoxin GelMA-UV bioink, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink Alg(Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink Bone UV/365 nm
Sigma-Aldrich
TissueFab® bioink , Alg(Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink , (GelAlgHA)MA Vis/405 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -VIS/405nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink , Alg(Gel)ma -Vis/525 nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Laminin -UV/365 nm, low endotoxin
Sigma-Aldrich
TissueFab® GelAlg − LAP Bioink, low endotoxin, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink Bone, Vis/405 nm
Sigma-Aldrich
TissueFab® bioink Bone, UV/365 nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Laminin -Vis/405 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, Fibronectin-UV/365nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (Vis/405), low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (UV/365), low endotoxin
Sigma-Aldrich
TissueFab® bioink Bone Vis/405 nm