Merck
Preparation and Membrane Properties of Oxidized Ceramide Derivatives.
Langmuir : the ACS journal of surfaces and colloids (2017-12-13)
Takaaki Matsufuji, Masanao Kinoshita, Anna Möuts, J Peter Slotte, Nobuaki Matsumori
ABSTRACT
Ceramide is a bioactive lipid with important roles in several biological processes including cell proliferation and apoptosis. Although 3-ketoceramides that contain a keto group in place of the 3-OH group of ceramide occur naturally, ceramide derivatives oxidized at the primary 1-OH group have not been identified to date. To evaluate how the oxidative state of the 1-OH group affects the physical properties of membranes, we prepared novel ceramide derivatives in which the 1-OH group was oxidized to a carboxylic acid (PCerCOOH) or methylester (PCerCOOMe) and examined the rigidity of their monolayers and the formation of gel domains in palmitoyloleoylphosphatidylcholine (POPC) or sphingomyelin (SM) bilayers. As a result, PCerCOOH and PCerCOOMe exhibited membrane properties similar to those of native ceramide, although the deprotonated form of PCerCOOH, PCerCOO-, exhibited markedly lower rigidity and higher miscibility with POPC and SM. This was attributed to the electrostatic repulsion of the negative charge, which hampered the formation of the ceramide-enriched gel domain. The similarities in the properties of PCerCOOMe and ceramide revealed the potential to introduce various functional groups onto PCerCOOH via ester or amide linkages; therefore, these derivatives will also provide a new strategy for developing molecular probes, such as fluorescent ceramides, and inhibitors of ceramide-related enzymes.
Product Description
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
C16 Ceramide (d18:1/16:0), Avanti Research - A Croda Brand