Merck
  • Influence of intrathecal injection with dexmedetomidine on the behavioral ability and analgesic effects on rats with neuropathic pain and expression of protein kinase C in the spinal dorsal horn.
Influence of intrathecal injection with dexmedetomidine on the behavioral ability and analgesic effects on rats with neuropathic pain and expression of protein kinase C in the spinal dorsal horn.
Experimental and therapeutic medicine (2018-10-23)
Xiaoning Li, Wei Zhang
ABSTRACT
This study aimed to investigate the influence of intrathecal injection with dexmedetomidine on the behavioral ability and analgesic effects of rat models with chronic neuropathic pain. It also discusses the role that the expression of protein kinase C (PKC) in the spinal dorsal horn plays in the analgesic mechanism of dexmedetomidine. A total of 35 Sprague Dawley rats were selected. Five rats were randomly chosen as the blank control group and the rest were prepared as models with chronic constriction injury, which were divided into the model and the observation group (n=15, each group). The observation group was injected with 10 ml dexmedetomidine intrathecally, while the model group was injected with an equivalent amount of saline. The behavioral abilities were evaluated by cumulative and motor function score. In addition, mechanical withdrawal threshold (MWT) and thermal withdrawal latency (TWL) were applied for the detection of pain threshold. Immunohistochemistry staining method was used to test the positive expression of PKC, and western blot analysis was used for the detection of PKC quantitative expression levels. After initiation, the instant cumulative and motor function score of the model and observation groups increased, while the values of MWT and TWL decreased. After initiation, the positive expression of PKC in the spinal dorsal horn of the model and observation groups rose dramatically, and the quantitative expression level was also on the rise. The positive and quantitative expression levels in the observation group reduced gradually over time. Thus, the intrathecal injection of dexmedetomidine may improve the behavioral ability of rats with chronic neuralgia and reduce the degree of pain, which may be associated with the inhibition of the expression of PKC in the spinal dorsal horn.
Sigma-Aldrich
Anti-Mouse (λ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-Protein Kinase C (PKC) antibody produced in mouse, clone MC5, ascites fluid