Merck
  • Triggered release of molecules across droplet interface bilayer lipid membranes using photopolymerizable lipids.
Triggered release of molecules across droplet interface bilayer lipid membranes using photopolymerizable lipids.
Langmuir : the ACS journal of surfaces and colloids (2012-05-03)
S Punnamaraju, H You, A J Steckl
ABSTRACT
A combination of nonpolymerizable phospholipids (DPPC or DPhPC) and a smaller amount of cross-linking photopolymerizable phospholipids (23:2 DiynePC) is incorporated in an unsupported artificial lipid bilayer formed using the droplet interface bilayer (DIB) approach. The DIB is formed by contacting lipid monolayer-coated aqueous droplets against each other in a dodecane-lipid medium. Cross-linking of the photopolymerizable lipids incorporated in the DIB was obtained by exposure to UV-C radiation (254 nm), resulting in pore formation. The effect of cross-linking on the DIB properties was characterized optically by measuring the diffusion of selectively encapsulated dye molecules (calcein) from one droplet of the DIB to the other droplet. Changes in DIB conductivity due to UV-C exposure were investigated using current-voltage (I-V) measurements. The leakage of dye molecules across the DIB and the increase in DIB conductivity after UV-C exposure indicates the formation of membrane pores. The results indicate that the DIB approach offers a simple and flexible platform for studying phototriggered drug delivery systems in vitro.
Product Description
Avanti
23:2 Diyne PE [DC(8,9)PE], 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder
Pricing and availability is not currently available.
Avanti
23:2 Diyne PC [DC(8,9)PC], Avanti Research - A Croda Brand 870016P, powder
Pricing and availability is not currently available.
Avanti
16:0-23:2 Diyne PE, 1-palmitoyl-2-(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder
Pricing and availability is not currently available.
Avanti
16:0-23:2 Diyne PC, 1-palmitoyl-2-(10,12-tricosadiynoyl)-sn-glycero-3-phosphocholine, powder
Pricing and availability is not currently available.