Merck
  • Fibronectin induces ureteric bud cells branching and cellular cord and tubule formation.
Fibronectin induces ureteric bud cells branching and cellular cord and tubule formation.
Kidney international (2004-10-02)
Peng Ye, Samy L Habib, Jill M Ricono, Nam-Ho Kim, Goutam G Choudhury, Jeffrey L Barnes, Hanna E Abboud, Mazen Y Arar
ABSTRACT
The extracellular matrix (ECM) protein fibronectin is involved in several stages of embryogenesis. Fibronectin exerts its effect through interaction with cellular integrin and nonintegrin receptors. We investigated the effect of fibronectin on branching and tubulogenesis of ureteric bud cells in a three-dimensional gel culture system. Primary ureteric bud cells from mouse embryos at gestation 11 days (E11) were isolated and established in culture. Fibronectin and integrin subunits were localized using immunoperoxidase staining. In three-dimensional collagen type I gel culture of ureteric bud cell, fibronectin dose dependently induces cord and tubule formation. Both ureteric bud cells and ureteric bud branches in embryonic kidney express the same multiple integrin subunits that include beta(1), beta(3), alpha(3), alpha(4) and alpha(v). Embryonic kidneys examined at E12, E14, and E16 days of gestation express fibronectin in the undifferentiated mesenchyme especially next to ureteric bud branches and in the interstitium around glomerulotubular structures and blood vessels. Fibronectin expression was similar at the tips and stalks of branching ureteric bud. Fibronectin expression is maximum at E12 and decreases with advanced gestation. Cultured ureteric bud cells also express fibronectin. RGD peptides inhibit cord and tubular formation in the three-dimensional gel. Anti-alpha(3)beta(1) antibody partially inhibits fibronectin-induced cord and tubule formation. Hepatocyte growth factor (HGF), fibroblast growth factor (FGF), and glial cell line-derived neurotrophic factor (GDNF) induce ureteric bud cell cord formation in three-dimensional gel. The effects of growth factors are delayed and quantitatively less compared to the effect of fibronectin. Fibronectin induces ureteric bud cells branching and tubulogenesis through interaction with multiple integrin receptors. Cultured ureteric bud cells express fibronectin and the origin of fibronectin at mesenchyme-ureteric bud interface is likely both the metanephric mesenchyme and ureteric bud epithelium. Addition of individual neutralizing antibodies to beta(1), beta(3), alpha(3), alpha(4,)alpha(6) and alpha(v) integrin subunits does not block the effect of fibronectin. Only an antibody to alpha(3)beta(1) integrin substantially blocks the effect of fibronectin. Other mechanisms, including unidentified integrins, are likely involved in fibronectin-induced cord and tubule formation.
Sigma-Aldrich
Anti-Integrin alphaV Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Fibronectin, Cellular antibody produced in mouse, clone FN-3E2, ascites fluid