Merck
  • A search for ceramide binding proteins using bifunctional lipid analogs yields CERT-related protein StarD7.
A search for ceramide binding proteins using bifunctional lipid analogs yields CERT-related protein StarD7.
Journal of lipid research (2018-01-19)
Svenja Bockelmann, John G M Mina, Sergei Korneev, Dina G Hassan, Dagmar Müller, Angelika Hilderink, Hedwich C Vlieg, Reinout Raijmakers, Albert J R Heck, Per Haberkant, Joost C M Holthuis
ABSTRACT
Ceramides are central intermediates of sphingolipid metabolism with dual roles as mediators of cellular stress signaling and mitochondrial apoptosis. How ceramides exert their cytotoxic effects is unclear and their poor solubility in water hampers a search for specific protein interaction partners. Here, we report the application of a photoactivatable and clickable ceramide analog, pacCer, to identify ceramide binding proteins and unravel the structural basis by which these proteins recognize ceramide. Besides capturing ceramide transfer protein (CERT) from a complex proteome, our approach yielded CERT-related steroidogenic acute regulatory protein D7 (StarD7) as novel ceramide binding protein. Previous work revealed that StarD7 is required for efficient mitochondrial import of phosphatidylcholine (PC) and serves a critical role in mitochondrial function and morphology. Combining site-directed mutagenesis and photoaffinity labeling experiments, we demonstrate that the steroidogenic acute regulatory transfer domain of StarD7 harbors a common binding site for PC and ceramide. While StarD7 lacks robust ceramide transfer activity in vitro, we find that its ability to shuttle PC between model membranes is specifically affected by ceramides. Besides demonstrating the suitability of pacCer as a tool to hunt for ceramide binding proteins, our data point at StarD7 as a candidate effector protein by which ceramides may exert part of their mitochondria-mediated cytotoxic effects.
Product Description
Avanti
C18 Ceramide (d18:1/18:0), Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-Mitochondria Antibody, surface of intact mitochondria, clone 113-1, clone 113-1, Chemicon®, from mouse
Avanti
18:1 Lyso PC, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
18:1 Lyso PC, Avanti Research - A Croda Brand
Avanti
18:1 Lyso PE, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
18:1 Lyso PE, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PG, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PG, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
C16 Ceramide (d18:1/16:0), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
Lyso SM (d18:1), Avanti Research - A Croda Brand 860600P, powder
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, chloroform
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, neat oil
Avanti
18:1 Lactosyl PE, Avanti Research - A Croda Brand