Merck
  • Further optimization of the K-Cl cotransporter KCC2 antagonist ML077: development of a highly selective and more potent in vitro probe.
Further optimization of the K-Cl cotransporter KCC2 antagonist ML077: development of a highly selective and more potent in vitro probe.
Bioorganic & medicinal chemistry letters (2012-06-26)
Eric Delpire, Aleksandra Baranczak, Alex G Waterson, Kwangho Kim, Nathan Kett, Ryan D Morrison, J Scott Daniels, C David Weaver, Craig W Lindsley
ABSTRACT
Further chemical optimization of the MLSCN/MLPCN probe ML077 (KCC2 IC(50)=537 nM) proved to be challenging as the effort was characterized by steep SAR. However, a multi-dimensional iterative parallel synthesis approach proved productive. Herein we report the discovery and SAR of an improved novel antagonist (VU0463271) of the neuronal-specific potassium-chloride cotransporter 2 (KCC2), with an IC(50) of 61 nM and >100-fold selectivity versus the closely related Na-K-2Cl cotransporter 1 (NKCC1) and no activity in a larger panel of GPCRs, ion channels and transporters.
Sigma-Aldrich
VU0463271, ≥98% (HPLC)