Merck
  • Two-dimensional crystals of carboxysome shell proteins recapitulate the hexagonal packing of three-dimensional crystals.
Two-dimensional crystals of carboxysome shell proteins recapitulate the hexagonal packing of three-dimensional crystals.
Protein science : a publication of the Protein Society (2009-10-22)
Kelly A Dryden, Christopher S Crowley, Shiho Tanaka, Todd O Yeates, Mark Yeager
ABSTRACT
Bacterial microcompartments (BMCs) are large intracellular bodies that serve as simple organelles in many bacteria. They are proteinaceous structures composed of key enzymes encapsulated by a polyhedral protein shell. In previous studies, the organization of these large shells has been inferred from the conserved packing of the component shell proteins in two-dimensional (2D) layers within the context of three-dimensional (3D) crystals. Here, we show that well-ordered, 2D crystals of carboxysome shell proteins assemble spontaneously when His-tagged proteins bind to a monolayer of nickelated lipid molecules at an air-water interface. The molecular packing within the 2D crystals recapitulates the layered hexagonal sheets observed in 3D crystals. The results reinforce current models for the molecular design of BMC shells.
Product Description
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand