Merck
  • Structure-activity relationship and mechanism of action studies of manzamine analogues for the control of neuroinflammation and cerebral infections.
Structure-activity relationship and mechanism of action studies of manzamine analogues for the control of neuroinflammation and cerebral infections.
Journal of medicinal chemistry (2009-12-19)
Jiangnan Peng, Sucheta Kudrimoti, Sivaprakasam Prasanna, Srinivas Odde, Robert J Doerksen, Hari K Pennaka, Yeun-Mun Choo, Karumanchi V Rao, Babu L Tekwani, Vamsi Madgula, Shabana I Khan, Bin Wang, Alejandro M S Mayer, Melissa R Jacob, Lan Chun Tu, Jürg Gertsch, Mark T Hamann
ABSTRACT
Structure-activity relationship studies were carried out by chemical modification of manzamine A (1), 8-hydroxymanzamine A (2), manzamine F (14), and ircinal isolated from the sponge Acanthostrongylophora. The derived analogues were evaluated for antimalarial, antimicrobial, and antineuroinflammatory activities. Several modified products exhibited potent and improved in vitro antineuroinflammatory, antimicrobial, and antimalarial activity. 1 showed improved activity against malaria compared to chloroquine in both multi- and single-dose in vivo experiments. The significant antimalarial potential was revealed by a 100% cure rate of malaria in mice with one administration of 100 mg/kg of 1. The potent antineuroinflammatory activity of the manzamines will provide great benefit for the prevention and treatment of cerebral infections (e.g., Cryptococcus and Plasmodium). In addition, 1 was shown to permeate across the blood-brain barrier (BBB) in an in vitro model using a MDR-MDCK monolayer. Docking studies support that 2 binds to the ATP-noncompetitive pocket of glycogen synthesis kinase-3beta (GSK-3beta), which is a putative target of manzamines. On the basis of the results presented here, it will be possible to initiate rational drug design efforts around this natural product scaffold for the treatment of several different diseases.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
5-Methoxytryptamine, 97%
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
5-Methoxytryptamine, British Pharmacopoeia (BP) Reference Standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Melatonin Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material