Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • New quinoline derivatives: synthesis and investigation of antibacterial and antituberculosis properties.
New quinoline derivatives: synthesis and investigation of antibacterial and antituberculosis properties.
European journal of medicinal chemistry (2010-06-12)
Sumesh Eswaran, Airody Vasudeva Adhikari, Imran H Chowdhury, Nishith K Pal, K D Thomas
ABSTRACT
Four new series of quinoline derivatives were synthesized starting from 2-trifluoromethyl aniline through multi-step reactions. In the reaction sequence, substituted aniline was cyclized to 4-hydroxy quinoline 1, which was then transformed to 4-chloro-2,8-bis(trifluoromethyl)quinoline 2. The key scaffold 4-hydrazinyl-2,8-bis(trifluoromethyl)quinoline 3, obtained from the compound 2, was successfully converted to target quinoline derivatives, viz. hydrazones 4a-t, ureas 5a-e, thioureas 6a-c and pyrazoles 7a-d, in good yields. The newly synthesized title compounds were evaluated for their in vitro antibacterial activity against Escherichia coli, Staphylococcus aureus, Pseudomonas aeruginosa and Klebsiella pneumoniae (recultured) and antituberculosis activity against Mycobacterium tuberculosis H(37)Rv and MDR-TB. Preliminary results indicated that most of the hydrazone derivatives demonstrated very good antibacterial and antituberculosis activities while other derivatives showed moderate activity.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals