Merck
  • Complex electrophysiological remodeling in postinfarction ischemic heart failure.
Complex electrophysiological remodeling in postinfarction ischemic heart failure.
Proceedings of the National Academy of Sciences of the United States of America (2018-03-14)
Bence Hegyi, Julie Bossuyt, Leigh G Griffiths, Rafael Shimkunas, Zana Coulibaly, Zhong Jian, Kristin N Grimsrud, Claus S Sondergaard, Kenneth S Ginsburg, Nipavan Chiamvimonvat, Luiz Belardinelli, András Varró, Julius G Papp, Piero Pollesello, Jouko Levijoki, Leighton T Izu, W Douglas Boyd, Tamás Bányász, Donald M Bers, Ye Chen-Izu
ABSTRACT
Heart failure (HF) following myocardial infarction (MI) is associated with high incidence of cardiac arrhythmias. Development of therapeutic strategy requires detailed understanding of electrophysiological remodeling. However, changes of ionic currents in ischemic HF remain incompletely understood, especially in translational large-animal models. Here, we systematically measure the major ionic currents in ventricular myocytes from the infarct border and remote zones in a porcine model of post-MI HF. We recorded eight ionic currents during the cell's action potential (AP) under physiologically relevant conditions using selfAP-clamp sequential dissection. Compared with healthy controls, HF-remote zone myocytes exhibited increased late Na+ current, Ca2+-activated K+ current, Ca2+-activated Cl- current, decreased rapid delayed rectifier K+ current, and altered Na+/Ca2+ exchange current profile. In HF-border zone myocytes, the above changes also occurred but with additional decrease of L-type Ca2+ current, decrease of inward rectifier K+ current, and Ca2+ release-dependent delayed after-depolarizations. Our data reveal that the changes in any individual current are relatively small, but the integrated impacts shift the balance between the inward and outward currents to shorten AP in the border zone but prolong AP in the remote zone. This differential remodeling in post-MI HF increases the inhomogeneity of AP repolarization, which may enhance the arrhythmogenic substrate. Our comprehensive findings provide a mechanistic framework for understanding why single-channel blockers may fail to suppress arrhythmias, and highlight the need to consider the rich tableau and integration of many ionic currents in designing therapeutic strategies for treating arrhythmias in HF.
Sigma-Aldrich
HMR-1556, ≥98% (HPLC)