Merck
  • Sema4C/PlexinB2 signaling controls breast cancer cell growth, hormonal dependence and tumorigenic potential.
Sema4C/PlexinB2 signaling controls breast cancer cell growth, hormonal dependence and tumorigenic potential.
Cell death and differentiation (2018-03-21)
Sreeharsha Gurrapu, Emanuela Pupo, Giulia Franzolin, Letizia Lanzetti, Luca Tamagnone
ABSTRACT
Semaphorin 4C (Sema4C) expression in human breast cancers correlates with poor disease outcome. Surprisingly, upon knock-down of Sema4C or its receptor PlexinB2 in diverse mammary carcinoma cells (but not their normal counterparts), we observed dramatic growth inhibition associated with impairment of G2/M phase transition, cytokinesis defects and the onset of cell senescence. Mechanistically, we demonstrated a Sema4C/PlexinB2/LARG-dependent signaling cascade that is required to maintain critical RhoA-GTP levels in cancer cells. Interestingly, we also found that Sema4C upregulation in luminal-type breast cancer cells drives a dramatic phenotypic change, with disassembly of polarity complexes, mitotic spindle misorientation, cell-cell dissociation and increased migration and invasiveness. We found that this signaling cascade is dependent on the PlexinB2 effectors ErbB2 and RhoA-dependent kinases. Moreover, Sema4C-overexpressing luminal breast cancer cells upregulated the transcription factors Snail, Slug and SOX-2, and formed estrogen-independent metastatic tumors in mice. In sum, our data indicate that Sema4C/PlexinB2 signaling is essential for the growth of breast carcinoma cells, featuring a novel potential therapeutic target. In addition, elevated Sema4C expression enables indolent luminal-type tumors to become resistant to estrogen deprivation, invasive and metastatic in vivo, which could account for its association with a subset of human breast cancers with poor prognosis.
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Y16, ≥98% (HPLC)
Sigma-Aldrich
Lapatinib
Sigma-Aldrich
Anti-Partitioning-defective 3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
ANTI-ZO1(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution