Merck
  • Rat d-aspartate oxidase is more similar to the human enzyme than the mouse enzyme.
Rat d-aspartate oxidase is more similar to the human enzyme than the mouse enzyme.
Biochimica et biophysica acta (2018-01-03)
Masumi Katane, Hisashi Kuwabara, Kazuki Nakayama, Yasuaki Saitoh, Tetsuya Miyamoto, Masae Sekine, Hiroshi Homma
ABSTRACT
d-Aspartate oxidase (DDO) is a degradative enzyme that is stereospecific for the acidic amino acid d-aspartate, an endogenous agonist of the N-methyl-d-aspartate (NMDA) receptor. Dysregulation of NMDA receptor-mediated neurotransmission has been implicated in the onset of various neuropsychiatric disorders including schizophrenia, as well as chronic pain. Thus, appropriate regulation of d-aspartate is believed to be important for maintaining proper neural activity in the nervous system. Accordingly, much attention has been paid to the role(s) of DDO in the metabolism of d-aspartate in vivo, and the physiological functions of DDO have been actively investigated using experimental rats and mice. However, detailed characterisation of rat DDO has not yet been performed, and little is known about species-specific differences in the properties of mammalian DDOs. In this study, the structural and enzymatic properties of purified recombinant rat, mouse and human DDOs were examined and compared. The results showed that rat DDO is more similar to human DDO than to mouse DDO. This work provides useful insight into the use of rats as an experimental model for investigating the biological significance of human DDO and/or d-aspartate. This article is part of a Special Issue entitled: d-Amino acids: biology in the mirror, edited by Dr. Loredano Pollegioni, Dr. Jean-Pierre Mothet and Dr. Molla Gianluca.
Sigma-Aldrich
5-Aminopyridine-3-carboxylic acid, 97%