Merck
  • Chlorophenoxyacetic acid and chloropyridylphenylurea accelerate translocation of photoassimilates to parthenocarpic and seeded fruits of muskmelon (Cucumis melo).
Chlorophenoxyacetic acid and chloropyridylphenylurea accelerate translocation of photoassimilates to parthenocarpic and seeded fruits of muskmelon (Cucumis melo).
Journal of plant physiology (2010-12-21)
Xin-Xian Li, Fumiyuki Kobayashi, Hiromi Ikeura, Yasuyoshi Hayata
ABSTRACT
We compared the effect of p-chlorophenoxyacetic acid (p-CPA) and 1-(2-chloro-4-pyridyl)-3-phenylurea (CPPU) on parthenocarpic and seeded muskmelon (Cucumis melo) fruits in regards to fruit development and the transport of photoassimilates from leaves exposed to ¹⁴CO₂ to the developing fruits. Ten days after anthesis (DAA), the fresh weight, total ¹⁴C-radioactivity and contents of ¹⁴C-sucrose and ¹⁴C-fructose were higher in the CPPU-induced parthenocarpic fruits than in seeded fruits. However, at 35 DAA, fresh weight and sucrose content in mesocarp, placenta and empty seeds of the parthenocarpic fruits were lower than in seeded fruits. Also, total ¹⁴C-radioactivity and ¹⁴C-sugar content of the parthenocarpic fruits were lower as well as the translocation rate of ¹⁴C-photoassimilates into these fruits. Application of p-CPA to the parthenocarpic fruits at 10 and 25 DAA increased fresh weight and sugar content. Moreover, these treatments elevated the total ¹⁴C-radioactivity, ¹⁴C-sucrose content and the translocation rate of ¹⁴C-photoassimilates. The ¹⁴C-radioactivity along the translocation pathway from leaf to petiole, stem, lateral shoot and peduncle showed a declining pattern but dramatically increased again in the fruits. These results suggest that the fruit's sink strength was regulated by the seed and enhanced by the application of p-CPA.
Product Description
Supelco
Forchlorfenuron, PESTANAL®, analytical standard
Sigma-Aldrich
N-(2-Chloro-4-pyridyl)-N′-phenylurea, applicable for cell culture, BioReagent
Supelco
4-Chlorophenoxyacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chlorophenoxyacetic acid, BioReagent, suitable for plant cell culture, crystalline
Sigma-Aldrich
4-Chlorophenoxyacetic acid, ≥98.0% (T)