Merck
  • A disposable electrochemical sensor for the determination of indole-3-acetic acid based on poly(safranine T)-reduced graphene oxide nanocomposite.
A disposable electrochemical sensor for the determination of indole-3-acetic acid based on poly(safranine T)-reduced graphene oxide nanocomposite.
Talanta (2011-06-08)
Tian Gan, Chengguo Hu, Zilin Chen, Shengshui Hu
ABSTRACT
A disposable electrochemical sensor for the determination of indole-3-acetic acid (IAA) based on nanocomposites of reduced graphene oxide (rGO) and poly(safranine T) (PST) was reported. The sensor was prepared by coating a rGO film on a pre-anodized graphite electrode (AGE) through dipping-drying and electrodepositing a uniform PST layer on the rGO film. Scanning electron microscopic (SEM) and infrared spectroscopic (IR) characterizations indicated that PST-rGO formed a rough and crumpled composite film on AGE, which exhibited high sensitive response for the oxidation of IAA with 147-fold enhancement of the current signal compared with bare AGE. The voltammetric current has a good linear relationship with IAA concentration in the range 1.0×10(-7)-7.0×10(-6)M, with a low detection limit of 5.0×10(-8)M. This sensor has been applied to the determination of IAA in the extract samples of several plant leaves and the recoveries varied in the range of 97.71-103.43%.
Sigma-Aldrich
Safranin O, indicator (pH 0.3-1.0), for microscopy (Bact., Bot., Hist.)
Sigma-Aldrich
Safranin O, certified by the BSC
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Gram′s safranin solution, for microscopy