Merck
  • Regulation of IL-1 signaling through control of focal adhesion assembly.
Regulation of IL-1 signaling through control of focal adhesion assembly.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-02-07)
Qin Wang, Julie Delcorde, Tracy Tang, Gregory P Downey, Christopher A McCulloch
ABSTRACT
IL-1 signaling is adhesion-restricted in many cell types, but the mechanism that drives it is not defined. We screened for proteins recruited to nascent adhesions in IL-1-treated human fibroblasts with tandem mass tag-mass spectrometry. We used fibronectin bead preparations to enrich 10 actin-associated proteins. There was a 1.2 times log 2-fold enrichment of actin capping protein (ACP) at 30 min after IL-1 stimulation. Knockdown (KD) of ACP by siRNA reduced IL-1-induced ERK activation(by 56%, matrix metalloproteinase-3 (MMP-3) expression by 48%, and MMP-9 expression by 62% (in all reductions, P < 0.01). Confocal or structured illumination microscopy showed that ACP was diffused throughout the cytosol but strongly accumulated at the ruffled border of spreading cells. ACP colocalized with nascent paxillin- and vinculin-containing adhesions at the ruffled border, but not with mature adhesions in the center. ACP KD promoted the formation of large, stable adhesions. Immunoprecipitation and proximity ligation analysis showed that ACP was associated with the IL-1 signal transduction proteins myeloid differentiation factor 88 (MyD88) and IL-1 receptor-associated kinase (IRAK) at the ruffled border of the leading edge. IL-1-induced phospho-ERK and MyD88 or IRAK colocalized at the leading edge. We concluded that ACP is required for recruitment and function of IL-1 signaling complexes in nascent adhesions at the leading edge of the cell.-Wang, Q., Delcorde, J., Tang, T., Downey, G. P., McCulloch, C. A. Regulation of IL-1 signaling through control of focal adhesion assembly.
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
MISSION® esiRNA, targeting human CAPZB
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit