Merck
  • Self-renewal versus lineage commitment of embryonic stem cells: protein kinase C signaling shifts the balance.
Self-renewal versus lineage commitment of embryonic stem cells: protein kinase C signaling shifts the balance.
Stem cells (Dayton, Ohio) (2011-02-11)
Debasree Dutta, Soma Ray, Pratik Home, Melissa Larson, Michael W Wolfe, Soumen Paul
ABSTRACT
The intricate molecular mechanisms that regulate ESC pluripotency are incompletely understood. Prior research indicated that activation of the Janus kinase-signal transducer and activator of transcription (STAT3) pathway or inhibition of extracellular signal-regulated kinase/glycogen synthase kinase 3 (ERK/GSK3) signaling maintains mouse ESC (mESC) pluripotency. Here, we demonstrate that inhibition of protein kinase C (PKC) isoforms maintains mESC pluripotency without the activation of STAT3 or inhibition of ERK/GSK3 signaling pathways. Our analyses revealed that the atypical PKC isoform, PKCζ plays an important role in inducing lineage commitment in mESCs through a PKCζ-nuclear factor kappa-light-chain-enhancer of activated B cells signaling axis. Furthermore, inhibition of PKC isoforms permits derivation of germline-competent ESCs from mouse blastocysts and also facilitates reprogramming of mouse embryonic fibroblasts toward induced pluripotent stem cells. Our results indicate that PKC signaling is critical to balancing ESC self-renewal and lineage commitment.
Sigma-Aldrich
Gö 6983, ≥97%
Sigma-Aldrich
MEK1/2 Inhibitor III, The MEK1/2 Inhibitor III, also referenced under CAS 391210-10-9, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Ro-31-8425, A potent, cell-permeable, reversible, ATP-competitive, and selective protein kinase C inhibitor (IC₅₀ = 15 nM for rat brain PKC).
Sigma-Aldrich
Gö 6983, A potent, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (PKC) that inhibits several PKC isozymes.
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)