Merck
  • Determination of volatile organic compounds in workplace air by multisorbent adsorption/thermal desorption-GC/MS.
Determination of volatile organic compounds in workplace air by multisorbent adsorption/thermal desorption-GC/MS.
Chemosphere (2004-04-28)
Chien-Hou Wu, Chien-Tai Feng, Yu-Shiu Lo, Tsai-Yin Lin, Jiunn-Guang Lo
ABSTRACT
Investigation of volatile organic compounds (VOCs) was first conducted in the air of class-100 cleanrooms at liquid crystal display (LCD) fabrication facilities. Air samples were collected on multisorbent tubes (including Carbopack B, Carbopack C, and Carbosieve S-III) and analyzed using adsorption/thermal desorption coupled with gas chromatography-mass spectrometry (GC-MS). Optimal conditions lead to average recoveries in the range of 96.2-98.2%, and method detection limits between 0.38 and 0.78 ppb, under the condition of 1-l sampling volume and 80% relative humidity. The method appears to be accurate, sensitive, simple and well-suited for determining VOC distributions from various stages of LCD manufacturing process and temporal variations of the analyte concentrations. About 15 VOCs were identified in workplace air. The major pollutants such as propylene glycol methyl ether acetate (PGMEA), butyl acetate, and acetone that are commonly used in the opto-electronics industry were detected and accurately quantified with the established method.
Product Description
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea