Merck
  • Application of thermal desorption to the development of a gas chromatographic/mass spectrometric method for the determination of toluene, chlorinated aromatic hydrocarbons, and 2,3,7,8-tetrachlorodibenzo-p-dioxin in combustion emissions.
Application of thermal desorption to the development of a gas chromatographic/mass spectrometric method for the determination of toluene, chlorinated aromatic hydrocarbons, and 2,3,7,8-tetrachlorodibenzo-p-dioxin in combustion emissions.
Journal of AOAC International (2003-02-28)
John D Donaldson, Susan M Grimes, Lina Mehta, Ahmad J Jafari
ABSTRACT
A fast and accurate analytical method, which uses commercially available adsorbents (Tenax TA, Carbotrap B and C, and Carbosieve S-III), was developed for the sampling and determination of aromatic hydrocarbons, chloroaromatic compounds, and 2,3,7,8-tetrachlorodibenzo-p-dioxin. The breakthrough volume data show that Carbotrap C has a good capacity for compounds of high molecular weight, whereas Carbosieve S-III and Tenax TA are efficient for volatile compounds. The organic components are thermally desorbed and transferred to a gas chromatograph/mass spectrometer. Importantly, thermal desorption avoids conventional solvent extraction procedures and also allows reuse of adsorbent tubes. Preliminary results for recovery of analytes from tubes packed with single adsorbent prove that a single-adsorbent bed is not capable of sampling a wide range of compounds. The best method to obtain the desired collection and desorption properties is to use adsorbent tubes containing several different materials. The results of optimization studies are summarized.
Product Description
Supelco
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 202, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Supelco
ORBO 101 Carbotrap® B (20/40), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 70 mm, pkg of 25 ea
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
ORBO 100 on Carbotrap® B (20/40), 350/175 mg, W,W,W separators, O.D. × L 7 mm × 110 mm, pkg of 25 ea
Supelco
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 500 g
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, stainless steel TD tube, unconditioned, pkg of 10 ea
Supelco
Carbotrap® 349
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea