Skip to Content
Merck
Zirconyl chloride octahydrate reagent grade, 98%

Zirconyl chloride octahydrate

Zirconyl chloride octahydrate reagent grade, 98%
Synonym(s):
Zirconium(IV) oxide chloride, Zirconium(IV) oxychloride octahydrate
Linear Formula:
ZrOCl2 · 8H2O
CAS Number:
Molecular Weight:
322.25
EC Number:

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing