
Ytterbium(III) trifluoromethanesulfonate hydrate

Synonym(s):
Trifluoromethanesulfonic acid ytterbium salt, Ytterbium(III) triflate
Linear Formula:
(CF3SO3)3Yb · xH2O
CAS Number:
Molecular Weight:
620.25 (anhydrous basis)
Products
Brand
Formula Weight
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing