
Withanoside V

Synonym(s):
(1α,3β,22R)-3-[(6-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]-1,22-dihydroxy-ergosta-5,24-dien-26-oic acid δ-lactone
Empirical Formula (Hill Notation):
C40H62O14
CAS Number:
Molecular Weight:
766.91
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing