Vinpocetine Related Compound B
Synonym(s):
Methyl (13aS,13bS)-13a-ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate, Apovincamine
Empirical Formula (Hill Notation):
C21H24N2O2
CAS Number:
Molecular Weight:
336.43
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2