
Vigabatrin impurity D

Synonym(s):
γ-Aminobutyric acid, 3-Carboxypropylamine, 4-Aminobutanoic acid, GABA, Piperidic acid, Piperidinic acid
Linear Formula:
NH2(CH2)3COOH
CAS Number:
Molecular Weight:
103.12
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing