Tritricosanoin
Synonym(s):
Glyceryl tritricosanoate, 1,2,3-Tritricosanoylglycerol, Tritricosanoin
Empirical Formula (Hill Notation):
C72H140O6
CAS Number:
Molecular Weight:
1101.88
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing