Skip to Content
Merck
Triton™ X-405 solution 70% in H2O

Triton X-405 solution

Triton™ X-405 solution 70% in H2O
Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol solution, Polyethylene glycol tert-octylphenyl ether, Polyoxyethylene (40) isooctylphenyl ether
CAS Number:

Products

Product Category
Feature
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing