Skip to Content
Merck
Tris[3-(trimethoxysilyl)propyl] isocyanurate technical grade

Tris[3-(trimethoxysilyl)propyl] isocyanurate

Tris[3-(trimethoxysilyl)propyl] isocyanurate technical grade
Empirical Formula (Hill Notation):
C21H45N3O12Si3
CAS Number:
Molecular Weight:
615.85

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing