
Triphenylphosphine-d15

Synonym(s):
Tri(phenyl-d5)phosphine
Linear Formula:
(C6D5)3P
CAS Number:
Molecular Weight:
277.38
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing