Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Triphenyl phosphate solution
Linear Formula:
(C
6
H
5
O)
3
PO
Molecular Weight:
326.28
Products
Product Category
stable isotopes (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Physical Form
Format
Markush Class
Markush Group
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
708968
Triphenyl phosphate solution,
NMR reference standard, 0.0485 M in acetone-d
6
(99.9 atom % D)
Expand
View Pricing
612863
Triphenyl phosphate solution,
NMR reference standard, 0.0485 M in chloroform-d (99.8 atom % D), NMR tube size 8 mm × 8 in.
Expand
View Pricing