Skip to Content
Merck
(Triisopropylsilyl)acetylene 97%

(Triisopropylsilyl)acetylene

(Triisopropylsilyl)acetylene 97%
Synonym(s):
Ethynyltriisopropylsilane
Linear Formula:
[(CH3)2CH]3SiC≡CH
CAS Number:
Molecular Weight:
182.38

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing