Skip to Content
Merck
Triethylene glycol di(p-toluenesulfonate) 98%

Triethylene glycol di(p-toluenesulfonate)

Triethylene glycol di(p-toluenesulfonate) 98%
Synonym(s):
2,2′-(Ethylenedioxy)diethyl ditosylate, Triethylene glycol ditosylate
Linear Formula:
(CH3C6H4SO3CH2CH2OCH2-)2
CAS Number:
Molecular Weight:
458.55

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing