Skip to Content
Merck
Tributylphenylstannane 97%

Tributylphenylstannane

Tributylphenylstannane 97%
Synonym(s):
Phenyltributylstannane, Phenyltributyltin, Tributylphenyltin
Linear Formula:
C6H5Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
367.16

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing