
Tri(2-furyl)phosphine

Synonym(s):
TFP, Tri-2-furanylphosphine, Tris(2-furanyl)phosphine, Tris(o-furyl)phosphine
Empirical Formula (Hill Notation):
C12H9O3P
CAS Number:
Molecular Weight:
232.17
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing