![Trehalose dihydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/172/189/26fde971-eadf-4d65-aaea-4cebeb282c55/640/26fde971-eadf-4d65-aaea-4cebeb282c55.png)
Trehalose, Dihydrate
![Trehalose dihydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/172/189/26fde971-eadf-4d65-aaea-4cebeb282c55/640/26fde971-eadf-4d65-aaea-4cebeb282c55.png)
Synonym(s):
Trehalose, Dihydrate, D-(+)-Trehalose dihydrate, α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside
Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing