Skip to Content
Merck
trans-2,3-Epoxybutane

trans-2,3-Epoxybutane

trans-2,3-Epoxybutane
Synonym(s):
trans-β-Butylene oxide, trans-2,3-Dimethyloxirane, trans-2-Butene oxide
Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing