Skip to Content
Merck
Trandolapril European Pharmacopoeia (EP) Reference Standard

Trandolapril

Trandolapril European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(2S,3aR,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid, Mavik, Trandolapril
Empirical Formula (Hill Notation):
C24H34N2O5
CAS Number:
Molecular Weight:
430.54

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade