Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trace elements in glass
Products
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (7)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
NIST610
Trace elements in glass,
NIST
®
SRM
®
610
Expand
View Pricing
NIST612
Trace elements in glass,
NIST
®
SRM
®
612
Expand
View Pricing
NIST614
Trace elements in glass,
NIST
®
SRM
®
614
Expand
View Pricing
NIST616
Trace elements in glass,
NIST
®
SRM
®
616
Expand
View Pricing
NIST617
Trace elements in glass,
NIST SRM 617
Expand
View Pricing
NIST611
Trace elements in glass,
NIST
®
SRM
®
611
Expand
View Pricing
NIST613
Trace elements in glass,
NIST
®
SRM
®
613
Expand
View Pricing