Tizanidine Related Compound B
Synonym(s):
1-Acetyl-N-(5-chloro-2,1,3-benzothiadiazol-4-yl)-4,5-dihydro-1H-imidazol-2-amine, N-Acetyltizanidine
Empirical Formula (Hill Notation):
C11H10ClN5OS
CAS Number:
Molecular Weight:
295.75
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2