Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5
Products
Manufacturer
Physical Form
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF60030649
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5,
foil, 1m coil, 0.030mm thickness, annealed
Expand
View Pricing
GF62639163
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5,
foil, 150 x 150mm, 0.030mm thickness, annealed, not light tested (nlt)
Expand
View Pricing
GF00515382
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5,
foil, 100 x 100mm, 0.030mm thickness, annealed, not light tested (nlt)
Expand
View Pricing
GF60061238
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5,
foil, 50 x 50mm, 0.030mm thickness, annealed, not light tested (nlt)
Expand
View Pricing
GF99237291
Titanium-aluminum-vanadium alloy, Ti94.5Al3V2.5,
foil, 0.5m coil, 0.030mm thickness, annealed
Expand
View Pricing