Tetrabutylammonium trifluoromethanesulfonate
Synonym(s):
Tetrabutylammonium triflate, Trifluoromethanesulfonic acid tetrabutylammonium salt
Linear Formula:
(CH3CH2CH2CH2)4N(CF3SO3)
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing