Skip to Content
Merck
Tetrabutylammonium bisulfate suitable for ion pair chromatography, LiChropur™, ≥99.0%

Tetrabutylammonium bisulfate

Tetrabutylammonium bisulfate suitable for ion pair chromatography, LiChropur™, ≥99.0%
Synonym(s):
Tetrabutylammonium hydrogen sulfate
Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing